Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
46.00 EUR - 660.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15560592
|
Thermo Scientific™
840-278000 |
This snap-on thermal printer attaches easily anddirectly to the back of the instrument and uses 80mm wide thermal drop in paper rolls. It can also be used with regular or self stick printer paper. |
660.00 EUR
Each |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15681465
|
Thermo Scientific™
840-259800 |
Self Stick Printer Paper |
125.00 EUR
10 rolls |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15551182
|
Thermo Scientific™
840-278300 |
Standard Printer Paper |
66.25 EUR
Pack of 10 |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15500602
|
Thermo Scientific™
840-278400 |
Transparent plastic dust cover protects instrument against dust and any liquid exposure while not in use. |
46.00 EUR
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15530602
|
Thermo Scientific™
840-278700 |
Pre-aligned, pre-assembled replacement Tungsten-Halogen lamp. Mouse-hole replacement access. Cover removal not required. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15510602
|
Thermo Scientific™
840-278500 |
Replacement Sample Compartment Lid. User-installed with basic tools. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Currently Unavailable: We have no information when this product will be available
|
Description
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible In addition to valuable built-in features such as the high resolution color display, large sample compartment, and data export options, a complete selection of accessories are available for the the GENESYS 30 spectrophotometer to help meet the needs of students, educators and lab technicians for accurate quantitative analysis.- Holds one rectangular longpath cell 20 - 100mm pathlength
- Comes assembled in a removable sample compartment liner
- Allows for flexibility between samples