Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Product Code. 15510602 Shop All Thermo Scientific Products
Click to view available options
Type:
Dust Cover
Replacement Compartment Lid
Self-Stick Printer Paper
Standard Printer Paper
Thermal Snap-on Printer
Tungsten Halogen Lamp
Unit Size:
10 rolls
Each
Pack of 10
Product Code. 15510602
Brand: Thermo Scientific™ 840278500
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Standard sample compartment lid
User installable with basic tools
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Compartment Lid
For Use With (Application) Replace your spectrophotometer lid and hinge assembly yourself if it is broken through abuse.
For Use With (Equipment) GENESYS 30 Visible Spectrophotometer
Includes 1 lid and hinge assembly
Storage Requirements Ambient
Warranty 2 Years