missing translation for 'onlineSavingsMsg'
Learn More

Gemigliptin, MedChemExpress

Product Code. p-200098155
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16482367

Brand: Medchem Express HY14892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gemigliptin (LC15-0444 ) is a highly selective, reversible and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. Gemigliptin exhibits potent anti-glycation properties. Gemigliptin can be used for the research of advanced glycation end products (AGE)-related diabetic complications.

Gemigliptin (LC15-0444 ) is a highly selective, reversible and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. Gemigliptin exhibits potent anti-glycation properties. Gemigliptin can be used for the research of advanced glycation end products (AGE)-related diabetic complications.

Specifications

Chemical Name or Material Gemigliptin
CAS 911637-19-9
Color Orange
Purity Grade Notes Research
Molecular Formula C18H19F8N5O2
Quantity 50 mg
Synonym LC15-0444
Solubility Information DMSO : 50 mg/mL (102.17 mM; Need ultrasonic)
SMILES O=C1N(C[C@@H](N)CC(N2CCC3=C(C(F)(F)F)N=C(C(F)(F)F)N=C3C2)=O)CC(F)(F)CC1
Molecular Weight (g/mol) 489.36
Formula Weight 489.36
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.