missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin/GSN Antibody (16H9), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 EUR - 533.00 EUR
Specifications
| Antigen | Gelsolin/GSN |
|---|---|
| Clone | 16H9 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043634
|
Novus Biologicals
NBP3-26541-50ul |
50 μL |
343.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043965
|
Novus Biologicals
NBP3-26541-100ul |
100 μL |
533.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gelsolin/GSN Monoclonal antibody specifically detects Gelsolin/GSN in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Gelsolin/GSN | |
| Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type) | |
| A synthesized peptide derived from Human Gelsolin/GSN [UniProt P06396] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 16H9 | |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2934 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title