Learn More
Gelatin, ( FCC), MP Biomedicals
Product Code. 11407251 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Product Code. 11407251
Brand: MP Biomedicals 0296031790
Chemical Identifiers
CAS 9000-70-8
MDL Number MFCD00081638
Specifications
Quantity 500 g
Grade FCC
Chemical Name or Material Gelatin, Type A; 100 Bloom