Learn More
STEMCELL Technologies Gel and PCR Clean-up Kit
Product Code. 17119911
Encompass_Preferred
Change view
Click to view available options
:
250 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17119911 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17119911 Supplier STEMCELL Technologies Supplier No. 79030
For extraction of DNA from agarose gels, or clean-up of PCR products and restriction enzyme digestions
  • The Gel and PCR Clean-up Kit uses a membrane-based system to quickly purify DNA fragments of 100 bp to 10 kb in length from polymerase chain reaction (PCR), restriction enzyme digestion, or excised agarose gel fragments (up to 350 mg)
  • The purified DNA is directly suitable for applications such as automated fluorescent DNA sequencing, cloning, labeling, restriction enzyme digestion, or in vitro transcription
  • Up to 40 μg of DNA can be processed per column in as little as 20 minutes and eluted in volumes as low as 15 μL, with up to 95% recovery depending on DNA fragment length
  • The Gel and PCR Clean-up Kit can be used to purify linear DNA fragments, supercoiled plasmid DNA, or single-stranded DNA (circular or linear)
Specifications
No. of Reactions 250 Preps