missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gefapixant, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Gefapixant is an orally active and potent purinergic P2X3 receptor (P2X3R) antagonist, with IC50 values of ∼30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Gefapixant can be used for the research of chronic cough and knee osteoarthritis.
Specifications
Specifications
| Chemical Name or Material | Gefapixant |
| CAS | 1015787-98-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H19N5O4S |
| Quantity | 25 mg |
| Synonym | MK-7264 AF-219 |
| Solubility Information | DMSO : 5 mg/mL (14.15 mM; ultrasonic and adjust pH to 5-6 with HCl) |
| SMILES | O=S(C1=CC(OC2=CN=C(N)N=C2N)=C(C(C)C)C=C1OC)(N)=O |
| Molecular Weight (g/mol) | 353.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction