missing translation for 'onlineSavingsMsg'
Learn More

Gefapixant, MedChemExpress

Product Code. 16409956
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409956

Brand: Medchem Express HY101588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gefapixant is an orally active and potent purinergic P2X3 receptor (P2X3R) antagonist, with IC50 values of ∼30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Gefapixant can be used for the research of chronic cough and knee osteoarthritis.

Gefapixant is an orally active and potent purinergic P2X3 receptor (P2X3R) antagonist, with IC50 values of ∼30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Gefapixant can be used for the research of chronic cough and knee osteoarthritis.

Specifications

Chemical Name or Material Gefapixant
CAS 1015787-98-0
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O4S
Quantity 50 mg
Synonym MK-7264 AF-219
Solubility Information DMSO : 5 mg/mL (14.15 mM; ultrasonic and adjust pH to 5-6 with HCl)
SMILES O=S(C1=CC(OC2=CN=C(N)N=C2N)=C(C(C)C)C=C1OC)(N)=O
Molecular Weight (g/mol) 353.4
Formula Weight 353.4
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.