missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDF7 Polyclonal Antibody

Product Code. 15558432
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15558432 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558432 Supplier Invitrogen™ Supplier No. PA547941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Approximately 10% cross-reactivity with recombinant mouse (rm) GDF-5 and rmGDF-6 is observed and less than 1% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, and recombinant human GDF-11 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF7
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene GDF7
Gene Accession No. P43029
Gene Alias BMP12; Gdf7; Gdf-7; growth differentiation factor 7; growth/differentiation factor 7
Gene Symbols GDF7
Host Species Goat
Immunogen E. coli-derived recombinant mouse GDF-7 Thr316-Arg461.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238057
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.