missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF3 Polyclonal Antibody
Description
Immunogen sequence: HKWIIAPKGF MANYCHGECP FSLTISLNSS NYAFMQALMH AVDPEIPQAV CIPTKLSPIS MLYQDNNDNV ILRHYEDMVV Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Specifications
Specifications
| Antigen | GDF3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GDF3 |
| Gene Accession No. | Q9NR23 |
| Gene Alias | C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 |
| Gene Symbols | GDF3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?