missing translation for 'onlineSavingsMsg'
Learn More

GDC-0575, MedChemExpress

Product Code. 16437307
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437307 5 mg 5mg
16484588 10 mg 10mg
16475436 100 mg 100mg
16470887 50 mg 50mg
16443146 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16437307 Supplier Medchem Express Supplier No. HY112167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0575 (ARRY-575, RG7741) is a highly-selective oral small-molecule Chk1 inhibitor with an IC50 of 1.2 nM.

GDC-0575 (ARRY-575, RG7741) is a highly-selective oral small-molecule Chk1 inhibitor with an IC50 of 1.2 nM.

Specifications

Chemical Name or Material GDC-0575
CAS 1196541-47-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H20BrN5O
Quantity 5 mg
Synonym ARRY-575 RG7741
Solubility Information DMSO : 100 mg/mL (264.36 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1CC1)NC2=CNC3=NC=C(Br)C(N4C[C@H](N)CCC4)=C32
Molecular Weight (g/mol) 378.27
Formula Weight 378.27
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.