missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 EUR - 550.00 EUR
Specifications
| Antigen | GCH1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232199
|
Novus Biologicals
NBP3-35157-20ul |
20 μL |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228949
|
Novus Biologicals
NBP3-35157-100ul |
100 μL |
550.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCH1 Polyclonal antibody specifically detects GCH1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| GCH1 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| dystonia 14, DYT14, DYT5a, DYT5EC 3.5.4.16, GCH, GTP cyclohydrolase 1, GTP cyclohydrolase I, GTPCH1, GTP-CH-1, GTP-CH-I, guanosine 5'-triphosphate cyclohydrolase I, HPABH4B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 55-250 of human GCH1 (NP_000152.1).,, Sequence:, GERPRSEEDNELNLPNLAAAYSSILSSLGENPQRQGLLKTPWRAASAMQFFTKGYQETISDVLNDAIFDEDHDEMVIVKDIDMFSMCEHHLVPFVGKVHIGYLPNKQVLGLSKLARIVEIYSRRLQVQERLTKQIAVAITEALRPAGVGVVVEATHMCMVMRGVQKMNSKTVTSTMLGVFREDPKTREEFLTLIRS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 2643 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title