missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GC Monoclonal Antibody (D4), Invitrogen™

Mouse Monoclonal Antibody
185.00 EUR - 415.00 EUR
Specifications
| Antigen | GC |
|---|---|
| Clone | D4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17970573
|
Invitrogen™
MA551072 |
20 μL |
185.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17959462
![]() |
Invitrogen™
MA551071 |
200 μL |
415.00 EUR
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Specifications
| GC | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor | |
| GC | |
| IgG1 κ | |
| Protein A/G | |
| GC |
| D4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02774 | |
| 2638 | |
| Recombinant Human DBP protein, Leu209-Asp401 (Accession #P02774). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title