Learn More
Macherey-Nagel™ GC Column Me-Sil Retention Gap
Product Code. 10398541 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
25 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Product Code. 10398541
Brand: Macherey-Nagel™ 723706.10
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type GC Column Retention Gap
Diameter (Metric) Inner 0.25 mm
For Use With (Application) n-hexane and Diethyl Ether Analysis
Temperature Range 320°C
Length (Metric) 10 m
Quantity 1 Each