missing translation for 'onlineSavingsMsg'
Learn More

GBR 12935 dihydrochloride, Thermo Scientific™

Product Code. 15485269
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
15485269 10 mg 10mg
15495269 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 15485269

Brand: Thermo Scientific Alfa Aesar J62991.MA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A very potent and selective inhibitor of dopamine uptake

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67469-81-2
Molecular Formula C28H36Cl2N2O
Molecular Weight (g/mol) 487.509
MDL Number MFCD00083175
InChI Key NQWRSILGEXNJIT-UHFFFAOYSA-N
Synonym gbr 12935 dihydrochloride, gbr-12935 dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxyethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-benzhydryloxyethyl-4-3-phenylpropyl piperazine;dihydrochloride, 1-2-benzhydryloxy ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-diphenylmethoxy ethyl-4-3-phenylpropyl piperazinediium dichloride, gbr12935 dihydrochloride
PubChem CID 11957553
ChEBI CHEBI:64091
IUPAC Name 1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;dihydrochloride
SMILES C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl

Specifications

Color White
Quantity 10 mg
Formula Weight 487.51
Physical Form Powder
Chemical Name or Material GBR 12935 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.