missing translation for 'onlineSavingsMsg'
Learn More

GBR 12935 dihydrochloride, MedChemExpress

Product Code. 16475038
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16475038 10 mg 10mg
16441277 50 mg 50mg
16405786 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 16475038

Brand: Medchem Express HY12242

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GBR 12935 dihydrochloride is a potent, and selective dopamine reuptake inhibitor.

GBR 12935 dihydrochloride is a potent, and selective dopamine reuptake inhibitor.

Specifications

Chemical Name or Material GBR 12935 dihydrochloride
Physical Form Solid
CAS 67469-81-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H36Cl2N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 25 mg/mL (51.28 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (14.65 mM; ultrasonic and warming and heat to 60°C)
SMILES N1(CCOC(C2=CC=CC=C2)C3=CC=CC=C3)CCN(CCCC4=CC=CC=C4)CC1.Cl.Cl
Molecular Weight (g/mol) 487.5
Formula Weight 487.5
Percent Purity 99.27%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.