missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA3 Monoclonal Antibody (OTI1C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502604S
This item is not returnable.
View return policy
Description
GBA3 Monoclonal antibody specifically detects GBA3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GBA3 | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H227 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57733 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1C7 | |
| Unconjugated | |
| GBA3 | |
| CBG; CBGL1; Cytosolic beta-glucosidase; cytosolic beta-glucosidase-like protein 1; Cytosolic GCase; Cytosolic glycosylceramidase; Gba3; GLUC; Glucosidase beta acid 3; glucosidase, beta, acid 3; glucosidase, beta, acid 3 (cytosolic); Glucosylceramidase beta 3; glucosylceramidase beta 3 (gene/pseudogene); klotho-related protein; KLRP | |
| Human recombinant protein fragment corresponding to amino acids 1-150 and 370-469 of human GBA3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction