missing translation for 'onlineSavingsMsg'
Learn More

GATAD1 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene

Product Code. 17534483
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17534483 100 μL 100µL
17554483 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17534483 Supplier Origene Technologies Supplier No. TA804787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Component of some chromatin complex recruited to chromatin sites methylated ′Lys-4′ of histone H3 (H3K4me).

Specifications

Antigen GATAD1
Applications Western Blot
Classification Monoclonal
Clone OTI2D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene GATAD1
Gene Accession No. Q8WUU5
Gene Alias CMD2B, ODAG, RG083M05.2
Gene Symbols GATAD1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 68-269 of human GATAD1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57798
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.