missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gastrin Polyclonal Antibody
GREENER_CHOICE

Product Code. 16653525
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16653525 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16653525 Supplier Invitrogen™ Supplier No. PA599275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Gastrin.

This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gastrin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gast
Gene Accession No. P01350, P04563, P48757
Gene Alias Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin
Gene Symbols Gast
Host Species Rabbit
Immunogen A synthesized peptide derived from human GAST(Accession P01350), corresponding to amino acid residues A70-N101.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14459, 2520, 25320
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.