Learn More
Lenz Laborglasintrumente™ Gas Washing Bottle Head with Filter Disc
Description
- Acc. to DIN 12596
- With cone NS 29/32
- Material: DURAN™ tubing
Specifications
Specifications
| Material | Duran Tubing |
| Type | GL 14 connections |
| Porosity | 1 |
| For Use With (Equipment) | Gas washing bottles acc. to DIN 12463 |
| Quantity | 1 |