Learn More
Trajan™ Gas-Tight Autosampler Syringes
Product Code. 10742002
Change view
Click to view available options
Volume (Metric):
5 μL
100 μL
Needle Gauge:
22 G
23 G
Connection Type:
Fixed
Removable
For Use With (Equipment):
CTC, Carlo Erba, Fisons Autosampler
Carlo Erba/Fisons/Leap/CTC ASD200 and AS800
PerkinElmer Autosystem
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Connection Type For Use With (Equipment) unitSize
10742002 5 μL 23 G Fixed PerkinElmer Autosystem Each
12997556 100 μL 22 G Removable Carlo Erba/Fisons/Leap/CTC ASD200 and AS800 Each
12977556 100 μL 22 G Fixed CTC, Carlo Erba, Fisons Autosampler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10742002 Supplier Trajan™ Supplier No. 001957
Only null left
Add to basket
Add to basket
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.02 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) 2.75 in.
Length (English) Needle 2.7 in.
Length (Metric) 70 mm
Product Type Autosampler Syringe
Model 50F-PE-0.63C
Quantity Ea.
Volume (Metric) 5 μL
Needle Point Style Cone
For Use With (Equipment) PerkinElmer Autosystem
Connection Type Fixed