Learn More
Marsh Bellofram Type 10 Air Pressure Regulator
Shop All Cole Parmer Products
Click to view available options
Material:
Brass
Unit Size:
Each
Description
- Use with clean, dry air.
- Ideal for gas mixing, valve operations, and calibration stands.
- These pressure regulators are designed for precise regulation of air pressure in pipes, vessels, and process lines.
- Regulators maintain constant pressure by providing high accuracy, precision control, and maximum stability under the most adverse conditions.
- Regulators feature high-accuracy pressure regulation up to ±0.1%, a servo-operated control mechanism for pressure stability, and start-up stability that returns to output setting after a long down-time.
- Locking nut prevents inadvertant adjustment of pressure.
- Unit can be in-line or panel mounted.
Specifications
Specifications
| Description | Pressure Regulator, 1/8 in. NPT(F), 2-25 psi |
| Type | Gas Regulator |
| For Use With (Equipment) | Cylinder |
| Material | Brass |