Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit, High Flow
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
Brand:  Trajan Scientific and Medical 1035004
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15720959
  • 540.00 EUR / Each
Estimated Shipment: 10-07-2024
Description
  • Easily installed into the gas line
  • Can be either wall-mounted or fixed to a bench
  • Available with a 1, 2 or 4 filter capacity
Specifications
Specifications
1/4 in. fittings
Gas filters
1/4 in.
2-position Connecting Unit High Flow
Each
Special Offers
For research use only