Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit, High Flow
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
520.00 EUR - 540.00 EUR
Specifications
Type | 2-position Connecting Unit High Flow |
---|---|
For Use With (Equipment) | Gas filters |
Packaging Quantity | Each |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
15730959
|
Trajan Scientific and Medical
1035008 |
1/8 in. |
520.00 EUR
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15720959
|
Trajan Scientific and Medical
1035004 |
1/4 in. |
540.00 EUR
Each |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Easily installed into the gas line
- Can be either wall-mounted or fixed to a bench
- Available with a 1, 2 or 4 filter capacity
Specifications
2-position Connecting Unit High Flow | |
Gas filters | |
Each |
For research use only