Learn More
Novus Biologicals™ Ganglioside GM3 Antibody (racotumomab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28885-100ug
 View more versions of this product
Product Code. 30048388
  • 326.00 EUR / 100µg
Description
Ganglioside GM3 Monoclonal antibody specifically detects Ganglioside GM3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ganglioside GM3
Monoclonal
Unconjugated
Q9UNP4
GM3
100 μg
Primary
Human
Purified
Flow Cytometry, ELISA, Functional Assay
racotumomab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Human
Protein A purified
RUO
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions