missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gandotinib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Gandotinib (LY2784544) is a potent JAK2 inhibitor with IC50 of 3 nM. Gandotinib (LY2784544) also inhibits FLT3, FLT4, FGFR2, TYK2, and TRKB with IC50 of 4, 25, 32, 44, and 95 nM.
Specifications
Specifications
| Chemical Name or Material | Gandotinib |
| CAS | 1229236-86-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25ClFN7O |
| Quantity | 50 mg |
| Synonym | LY2784544 |
| Solubility Information | DMSO : ≥ 50 mg/mL (106.40 mM) |
| SMILES | CC1=NNC(NC2=NN3C(C(CN4CCOCC4)=C2)=NC(C)=C3CC5=CC=C(C=C5F)Cl)=C1 |
| Molecular Weight (g/mol) | 469.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction