Learn More
Berkshire™ Gamma™ SuperSorb™ Low Endotoxin Wipe
BIOPHARMA
Product Code. 17161101 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
30 x 30 cm
Unit Size:
Pack of 160
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17161101 30 x 30 cm Pack of 160
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17161101 Supplier Berkshire™ Supplier No. GWSSST16LE
Only null left
Add to basket
Add to basket
Berkshire’s line of low endotoxin cleanroom wipes have been engineered to meet the specific requirements of the pharmaceutical, healthcare and medical device industries.
Specifications
Material Polyester
Construction Knitted
Edges Ultrasonically Sealed
Dimensions (L x W) 30 x 30 cm
Certifications/Compliance (Europe) guaranteed endotoxin limits - <20 EU/device - <0.5 EU/mL