Learn More
asecos™ Galvanised Steel Spray Can
Product Code. p-200003678 Shop All asecos Products
Click to view available options
Capacity (Metric):
1 L
2 L
Color:
Gray
Yellow
Unit Size:
Each
Product Code. 17856446
Brand: asecos™ 23137
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Galvanised Steel Spray Can is ideal for economical and controlled spraying of surfaces and parts to be cleaned.
  • Spray attachment made of plastic with adjustable nozzle.
  • Integrated plastic pump.
  • Incl. multilingual safety sticker to designate the contents.
Specifications
Product Type Spray Can
Capacity (English) 0.3 gal.
Capacity (Metric) 1 L
Color Yellow
For Use With (Application) Spray cleaning surfaces sparingly and in measured doses
Material Steel
Width (English) 6.38 in.
Width (Metric) 162.00 mm
Height (English) 11.81 in.
Height (Metric) 300.00 mm
Weight (English) 1.65 lb.
Weight (Metric) 0.75 kg