missing translation for 'onlineSavingsMsg'
Learn More

Gadobutrol, MedChemExpress

Product Code. 16487066
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487066 100 mg 100mg
16471216 200 mg 200mg
16457008 10 mg 10mg
16453057 50 mg 50mg
16409557 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487066 Supplier Medchem Express Supplier No. HY16217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gadobutrol (Gd-DO3A-butrol; ZK 135079) is a nonionic paramagnetic macrocyclic gadolinium-based contrast agent that can be used for magnetic resonance imaging (MRI).

Gadobutrol (Gd-DO3A-butrol

Specifications

Chemical Name or Material Gadobutrol
Physical Form Solid
CAS 770691-21-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H31GdN4O9
Synonym ZK 135079
Solubility Information H2O : 20 mg/mL (33.07 mM; Need ultrasonic)
SMILES OC[C@H]1[N]23[Gd+3]([N]4(C5)CC3)([N]6(C7)CC4)([OH][C@@H]1CO)([O-]C5=O)([O-]C8=O)([O-]C7=O)[N](CC6)(C8)CC2
Molecular Weight (g/mol) 604.71
Formula Weight 604.71
Percent Purity 99.96%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.