missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6PD Mouse anti-Human, Unconjugated, Clone: 2A7B12, Proteintech
Mouse Monoclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | G6PD |
|---|---|
| Clone | 2A7B12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818195
|
Proteintech
66373-1-IG-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808195
|
Proteintech
66373-1-IG-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The G6PD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human G6PD. This antibody recognizes human, mouse, rat antigen. The G6PD antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| G6PD | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 2539 | |
| Primary |
| 2A7B12 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title