Learn More
G 5555, Tocris Bioscience™
GREENER_CHOICE
High affinity group I PAK inhibitor
Brand:  Tocris Bioscience 6051/50
Encompass
Product Code. 15726359
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
G 5555 is a high affinity group 1 PAK (pan-PAK1, 2 and 3) inhibitor (Ki = 3.7 nM; pMEK IC50 = 69 nM). Of 235 kinases tested, other than PAK1, eight (PAK2, PAK3, KHS1, Lck, MST3, MST4, SIK2, and YSK1) were inhibited >70%. Exhibits low hERG channel activity (<50% inhibition at 10 μM). Orally bioavailable.
Specifications
Specifications
8-[(trans-5-Amino-1,3-dioxan-2-yl)methyl]-6-[2-chloro-4-(6-methyl-2-pyridinyl)phenyl]-2-(methylamino)-pyrido[2,3-d]pyrimidin-7(8H)-one
1648863-90-4
>98%
50 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions