missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD9 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547321
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 2% cross-reactivity with recombinant mouse (rm) Frizzled-2, rmFrizzled-3, rmFrizzled-4, rmFrizzled-6, rmFrizzled-7, and rmFrizzled-8 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
Specifications
| FZD9 | |
| Polyclonal | |
| Unconjugated | |
| FZD9 | |
| CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 14371 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9R216 | |
| FZD9 | |
| Sf 21-derived recombinant mouse Frizzled-9 Leu24-Asp186. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction