Learn More
A&D Weighing FX/FZ Precision Balances IP 65 Class II Approved
GREENER_CHOICE
Product Code. 16601092
Click to view available options
Capacity:
122 g
220 g
320 g
1220 g
2200 g
3200 g
Unit Size:
Each
Product Code. 16601092
Brand: A&D Weighing FZ1200iWPEC
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
  • IP65 water / dust proof
  • Built-in internal calibration at the touch of a button
  • Shock absorber function
  • Auto power ON/OFF function
  • B5 size footprint for use in narrow spaces
  • VFD Display for easy to read weighing results
  • SCF – Statistical Calculation Function
  • Standard RS-232C
  • Fast compact SHS sensor, response time only 1 second
  • Quick USB interface – no driver installation required (optional)
  • LAN – Ethernet interface with WinCT-Plus software (optional)
  • Animal weighing pan (optional)
  • Large breeze break (optional)
  • Small breeze break included on 1 mg models
  • Built-in rechargeable NiMH battery pack (optional)
Specifications
Certifications/Compliance GLP/GMP/GCP/ISO/IP65/OIML
Connection Type RS-232 C, Quick USB (option), LAN-Ethernet (option)
Capacity 1220 g
Display Type VFD
Readability 0.01 g
Material Stainless Steel
Model FZ-1200i-WP-EC
Stabilization Time 1 second
Electrical Requirements 11 VA to the adaptor (12 V, 1 A)