missing translation for 'onlineSavingsMsg'
Learn More

Fusidic Acid Sodium Salt, TRC

Product Code. 30008481
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008481 50 mg 50mg
30012000 100 mg 100mg
30008788 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008481 Supplier Toronto Research Chemicals Supplier No. TRCF86550550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 751-94-0
Molecular Formula C31 H47 O6 . Na
Molecular Weight (g/mol) 538.69
Synonym Sodium Fusidate, 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, sodium salt (1:1), (3α,4α,8α,9β,11α,13α,14β,16β,17Z)-, 29-Nor-8α,9β,13α,14β-dammara-17(20),24-dien-21-oic acid, 3α,11α,16β-trihydroxy-, 16-acetate, monosodium salt, (Z)- (8CI), 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3,11-dihydroxy-, monosodium salt, (3α,4α,8α,9β,11α,13α,14β,16β,17Z)- (9CI), Fucidin, Fucidina, Fucidine, Fusidate sodium, Fusidic acid sodium salt, Fusidin, SQ 16360, Sodium fucidate, Sodium fusidate, Sodium fusidin, ZN 6-Na, Sodium ent-(17Z)-16α-(acetyloxy)-3β,11β-dihydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oate
IUPAC Name sodium;(2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoate
SMILES [Na+].C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)[O-])\[C@H](C[C@]34C)OC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C31H48O6.Na/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);/q;+1/p-1/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1
Formula Weight 538.327
Percent Purity >95%
Chemical Name or Material Fusidic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.