Learn More
Furosemide Acyl-Beta-D-glucuronide, TRC
Product Code. 30009412
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009412 10 mg 10mg
30007601 5 mg 5mg
30011661 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009412 Supplier Toronto Research Chemicals Supplier No. TRCF86501010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72967-59-0
Molecular Formula C18 H19 Cl N2 O11 S
Molecular Weight (g/mol) 506.87
Synonym β-D-Glucopyranuronic acid, 1-[5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoate], Furosemide glucuronide, Furosemide Acyl-β-D-glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES NS(=O)(=O)c1cc(C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)c(NCc3occc3)cc1Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C18H19ClN2O11S/c19-9-5-10(21-6-7-2-1-3-30-7)8(4-11(9)33(20,28)29)17(27)32-18-14(24)12(22)13(23)15(31-18)16(25)26/h1-5,12-15,18,21-24H,6H2,(H,25,26)(H2,20,28,29)/t12-,13-,14+,15-,18-/m0/s1
Formula Weight 506.0398
Chemical Name or Material Furosemide Acyl-beta-D-glucuronide