missing translation for 'onlineSavingsMsg'
Learn More

Furazolidone D4 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314559
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314559 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314559 Supplier Dr. Ehrenstorfer Supplier No. DREA13970210AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furazolidone D4
CAS 1217222-76-8
Solvent or Matrix Acetonitrile
Molecular Formula C8 2H4 H3 N3 O5
Quantity 1 mL
Synonym 4,4,5,5-Tetradeuterio-3-[(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
InChI Formula InChI=1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+/i3D2,4D2
SMILES [2H]C1([2H])OC(=O)N(\N=C\c2oc(cc2)[N+](=O)[O-])C1([2H])[2H]
IUPAC Name 4,4,5,5-tetradeuterio-3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 229.1829
Formula Weight 229.0637 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.