Learn More
DWK Life Sciences DURAN™ Blank for separating and dropping funnel, cylindrical with stopper bed for ISO 4800
Shop All DWK Life Sciences Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
missing translation for 'unitSize'
Pack of 10
Description
DURAN™ dropping funnels are used, for example, for the metered supply of liquid to a reaction mixture.
- Cylindrical with stopper bed for ISO 4800
- Glass articles for further processing
Specifications
Specifications
| Diameter (Metric)Top | 85 mm |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass (3.3) |
| Autoclavable | Autoclavable |
| For Use With (Application) | Blank for further processing as dropping funnel; Phase separation |
| Quantity | 10/Pk. |