Learn More
Manta Diagnostics™ Funnel, Base Support and Clip
Click to view available options
Capacity (Metric):
300 mL
500 mL
1000 mL
Unit Size:
Each
Description
300 mL, 500 mL, and 1 L options available
Specifications
Specifications
| Capacity (Metric) | 300 mL |
| Material | Glass |
| Autoclavable | Autoclavable |
| Graduated | No |
| Quantity | 1 |