missing translation for 'onlineSavingsMsg'
Learn More

Fumagillin, TRC

Product Code. 30004671
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004671 5 mg 5mg
30007321 10 mg 10mg
30003562 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004671 Supplier Toronto Research Chemicals Supplier No. TRCF8626505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23110-15-8
Molecular Formula C26 H34 O7
Molecular Weight (g/mol) 458.54
Synonym 2,4,6,8-Decatetraenedioic acid, 1-[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methyl-2-buten-1-yl)-2-oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, (2E,4E,6E,8E)-, 2,4,6,8-Decatetraenedioic acid 4-(1,2-epoxy-1,5-dimethyl-4-hexenyl)-5-methoxy-1-oxaspiro[2.5]oct-6-yl ester (6CI), 2,4,6,8-Decatetraenedioic acid, mono[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methyl-2-butenyl)oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, (2E,4E,6E,8E)- (9CI), 2,4,6,8-Decatetraenedioic acid, mono[4-(1,2-epoxy-1,5-dimethyl-4-hexenyl)-5-methoxy-1-oxaspiro[2.5]oct-6-yl] ester (8CI), 2,4,6,8-Decatetraenedioic acid, mono[5-methoxy-4-[2-methyl-3-(3-methyl-2-butenyl)oxiranyl]-1-oxaspiro[2.5]oct-6-yl] ester, [3R-[3α,4α(2R*,3R*),5β,6β(2E,4E,6E,8E)]]-, 1-Oxaspiro[2.5]octane, 2,4,6,8-decatetraenedioic acid deriv., (-)-Fumagillin, Amebacilin, Fugillin, Fumadil B, Fumagillin, Fumagillin DCH, Fumidil, NSC 9168, ZGN 201
IUPAC Name (2E,4E,6E,8E)-10-[[(3R,4S,5S,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methylbut-2-enyl)oxiran-2-yl]-1-oxaspiro[2.5]octan-6-yl]oxy]-10-oxodeca-2,4,6,8-tetraenoic acid
SMILES CO[C@@H]1[C@@H](CC[C@]2(CO2)[C@H]1[C@@]3(C)O[C@@H]3CC=C(C)C)OC(=O)\C=C\C=C\C=C\C=C\C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H34O7/c1-18(2)13-14-20-25(3,33-20)24-23(30-4)19(15-16-26(24)17-31-26)32-22(29)12-10-8-6-5-7-9-11-21(27)28/h5-13,19-20,23-24H,14-17H2,1-4H3,(H,27,28)/b7-5+,8-6+,11-9+,12-10+/t19-,20-,23-,24-,25+,26+/m1/s1
Formula Weight 458.2305
Chemical Name or Material Fumagillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.