Learn More
Fumagillin, MedChemExpress
Product Code. 16488598
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488598 10 mg 10mg
16453796 25 mg 25mg
16452269 1 mg 1mg
16401058 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16488598 Supplier Medchem Express Supplier No. HYB0751
MedChemExpress Fumagillin(NSC9168) is an antimicrobial compound first isolated in 1949 from the fungus Aspergillus fumigatu. Fumagillin can inhibits HIV‐1 infection through the inhibition of HIV-1 viral protein R (Vpr) activity.
Fumagillin(NSC9168) is an antimicrobial compound first isolated in 1949 from the fungus Aspergillus fumigatu. Fumagillin can inhibits HIV‐1 infection through the inhibition of HIV-1 viral protein R (Vpr) activity.
Specifications
Chemical Name or Material Fumagillin
Physical Form Solid
CAS 23110-15-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H34O7
For Use With (Application) COVID-19-anti-virus
Synonym Amebacilin NSC9168
Solubility Information DMSO : 50 mg/mL (109.04 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]1([C@@H](C/C=C(C)/C)O1)[C@]([C@@H]2OC)([H])[C@]3(CC[C@H]2OC(/C=C/C=C/C=C/C=C/C(O)=O)=O)CO3
Molecular Weight (g/mol) 458.54
Formula Weight 458.54
Percent Purity 95.0%
Grade Research