Learn More
3M™ Fuel™ X2 Safety Glasses
Description
- Safety Glasses with a sleek half-frame profile helps ensure that protection and performance remain uncompromised
- The frame design has bold new grooved styling and a fully adjustable nose bridge
- Adjustable temple length for comfort and optimal fit
- Optical class 1 for maximum clarity
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Frame made from a high density nylon material for impact, torsion and heat resistance
- special mirror coating provides glare reduction and UV protection
- Strong polycarbonate lens for impact resistance
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 166:2001 |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Bronze |
| Product Type | Safety glasses |
| Frame Color | Brown |
| Lens Material | Polycarbonate |