missing translation for 'onlineSavingsMsg'
Learn More

FUBP3 Polyclonal Antibody

Product Code. 16539711
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16539711 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16539711 Supplier Invitrogen Supplier No. PA596875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FUBP3 Polyclonal Antibody

FUBP3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FUBP3
Gene Accession No. Q96I24
Gene Alias A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2
Gene Symbols FUBP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 65-260 of human FUBP3 (NP_0039251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320267, 8939
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.