Learn More
FTI 276, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
FTI-276 is a protein farnesyl transferase (PFT) inhibitor with IC50s of 0.9 and 0.5 nM for Plasmodium falciparum and human.
Specifications
Specifications
| Chemical Name or Material | FTI 276 |
| CAS | 170006-72-1 |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27N3O3S2 |
| For Use With (Application) | Cancer-programmed cell death |
| SMILES | O=C(O)[C@@H](NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=CC=CC=C2)=O)CCSC |
| Molecular Weight (g/mol) | 433.59 |
| Formula Weight | 433.59 |
| Grade | Research |