missing translation for 'onlineSavingsMsg'
Learn More

FT827, MedChemExpress

Product Code. p-200098287
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480737

Brand: Medchem Express HY111350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress FT827 is a selective and covalent ubiquitin-specific protease 7 (USP7) inhibitor (Ki=4.2 μM). FT827 binds to the USP7 catalytic domain (USP7CD; residues 208-560) with an apparent Kd value of 7.8 μM.

FT827 is a selective and covalent ubiquitin-specific protease 7 (USP7) inhibitor (Ki=4.2 μM). FT827 binds to the USP7 catalytic domain (USP7CD

Specifications

Chemical Name or Material FT827
CAS 1959537-86-0
Color White
Purity Grade Notes Research
Molecular Formula C27H28N6O5S
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (227.85 mM; Need ultrasonic)
SMILES C=CS(=O)(NC1=CC=CC=C1C2=CC=C(C(N3CCC(O)(CN(C=NC4=C5C=NN4C)C5=O)CC3)=O)C=C2)=O
Molecular Weight (g/mol) 548.61
Formula Weight 548.61
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.