Learn More
Vileda Professional™ Swep Duo Plus Mop Frame
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Pack of 10
Description
- Built-in lever designed to allow hands free removal of used mop heads
- Double sided that works with all core and specialist application
- Reinforced swivel joint part
- Trapezoidal shape for easy cleaning of corners
- Attaches to extendable handles (Recommended Aluminum telescopic handle - 119966)
- Lightweight and ergonomic
Specifications
Specifications
| Product Type | Mop Frame |
| Autoclavable | Autoclavable |
| Size | 50 cm (19.7 in.) |
| For Use With (Equipment) | Telescopic Handle, Aluminum Handle |
| For Use With (Application) | Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs |