Learn More
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7
Product Code. 15733245
Product Code. 15733245 Supplier Eppendorf™ Supplier No. 6001021310
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F740 series (5-compartment, compartment 1-4), 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 167 (22.4 x 5.5 x 6.6 in)- Max. box number per rack: 8 (4 x 2)- Stainless steel- For ULT freezer: Premium U410 (max. 15 racks), Premium U700 (max. 25 racks), HEF™ U410 (max. 15 racks), CryoCube™ F570 series (max. 25 racks), CryoCube F740 series (5-compartment version, compartment 1-4)- For ULT freezer: Premium U410 (max. 120 boxes), Premium U700 (max. 200 boxes), HEF™ U410 (max. 120 boxes), CryoCube™ F570 series (max. 200 boxes), CryoCube F740 series (5-compartment version, compartment 1-4)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack