Learn More
Eppendorf™ Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F7
Description
Freezer Rack: Premium U410, U700, HEF™ U410, CryoCube™ F570 series, CryoCube™ F740 series (5-compartment, compartment 1-4), 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 167 (22.4 x 5.5 x 6.6 in)- Max. box number per rack: 8 (4 x 2)- Stainless steel- For ULT freezer: Premium U410 (max. 15 racks), Premium U700 (max. 25 racks), HEF™ U410 (max. 15 racks), CryoCube™ F570 series (max. 25 racks), CryoCube F740 series (5-compartment version, compartment 1-4)- For ULT freezer: Premium U410 (max. 120 boxes), Premium U700 (max. 200 boxes), HEF™ U410 (max. 120 boxes), CryoCube™ F570 series (max. 200 boxes), CryoCube F740 series (5-compartment version, compartment 1-4)
Specifications
Specifications
| Product Type | Freezer Rack |