Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, DWP (53 mm)
Description
Freezer Rack: Innova™ C580, C760, locking rod, DWP (53 mm), side access, stainless steel- Rack for DWP (53 mm/ 2 in) - Max. box size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 140 x 91 x 776 (5.5 x 3.6 x 30.6 in)- Max. box number per rack: 14 (14 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 52 racks), Innova C760 (max. 66 racks)- For ULT freezer: Innova C580 (max. 728 DWP), Innova C760 (max. 924 DWP)- With locking rod for increased sample safety
Specifications
Specifications
| Product Type | Freezer Rack |