Learn More
Tenak™ Freezer Rack with Divider
Shop All TENAK Products
Click to view available options
Holds:
2 x 4 Goblets
3 x 4 Goblets
4 x 4 Goblets
Unit Size:
Each
Description
- Includes divider
- For use with freezer
Specifications
Specifications
| Dimensions (L x W x H) | 610 x 140 x 140 mm |
| For Use With (Equipment) | Freezers |
| Holds | 4 x 4 Goblets |
| Design | With Divider |
| Type | Rack |