Learn More
Eppendorf™ Freezer Rack: CryoCube™ F740 series (5-comp., MAX for 5th comp.), 2 in/53 mm
Product Code. 16387105
Change view
Click to view available options
Dimensions (L x W x H):
56.9 x 13.9 x 34.3 cm/5.5 x 22.4 x 13.5 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
16387105 56.9 x 13.9 x 34.3 cm/5.5 x 22.4 x 13.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16387105 Supplier Eppendorf™ Supplier No. 6001081210
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: CryoCube™ F740 series (5-compartment, MAX for 5th comp.), 2 in/53 mm, side access, stainless steel- Rack for 53 mm/ 2.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack (MAX) (5th compartment)- Ext. dimensions of rack (DxWxH) [mm]: 563 x 140 x 346 (22.2 x 5.5 x 13.6 in)- Max. box number per rack: 24 (4 x 6)- Material: Stainless steel- For ULT freezer: CryoCube F740 series 5-compartment (6x MAX racks in 5th compartment, 24x racks in compartment 1-4, all compartments: 30x racks)- For ULT freezer: CryoCube F740 series 5-compartment (Max. 144x boxes in 5th compartment, 384x boxes in compartment 1-4, all compartments: 528x boxes)
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 56.9 x 13.9 x 34.3 cm/5.5 x 22.4 x 13.5 in.
Product Type Freezer Rack