Learn More
Hanna Instruments™ Free and Total Chlorine Portable Photometer
Product Code. 11376360
Change view
Click to view available options
Range:
0 to 5mg/L
Length (Metric):
192 mm
193 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Length (Metric) unitSize
11376360 0 to 5mg/L 193 mm Each
11667130 0 to 5mg/L 192 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11376360 Supplier Hanna Instruments™ Supplier No. HI96711
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Portable Photometer
Accuracy ±0.03mg/L
For Use With (Application) Free and Total Chlorine
Height (Metric) 69 mm
Length (Metric) 193 mm
Light Source Tungsten Lamp
Range 0 to 5mg/L
Resolution 0.01mg/L
Weight (Metric) 0.36 kg
Width (Metric) 104 mm